accessibility menu, dialog, popup

UserName

H3COOCH3CH3

CAS RN 25263-97-2

IUPAC Name: 2-methylpropyl tetradecanoate
Synonyms: isobutyl myristate
Molecular Weight (g/mol): 284.48
Molecular Formula: C18H36O2
InChi Key: SKVCWXRLKHBEKW-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)OCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results