missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl Methacrylate (stabilized with HQ) 98.0+%, TCI America™
$39.14 - $67.11
Chemical Identifiers
| CAS | 97-86-9 |
|---|---|
| Molecular Formula | C8H14O2 |
| Molecular Weight (g/mol) | 142.20 |
| MDL Number | MFCD00008931,MFCD00084435 |
| InChI Key | RUMACXVDVNRZJZ-UHFFFAOYSA-N |
| Synonym | isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove |
| PubChem CID | 7352 |
| IUPAC Name | 2-methylpropyl 2-methylprop-2-enoate |
| SMILES | CC(C)COC(=O)C(C)=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M013225ML
|
TCI America
M013225ML |
25 mL |
Each for $39.14
|
|
|||||
|
M0132500ML
|
TCI America
M0132500ML |
500 mL |
Each for $67.11
|
|
|||||
Chemical Identifiers
| 97-86-9 | |
| 142.20 | |
| RUMACXVDVNRZJZ-UHFFFAOYSA-N | |
| 7352 | |
| CC(C)COC(=O)C(C)=C |
| C8H14O2 | |
| MFCD00008931,MFCD00084435 | |
| isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove | |
| 2-methylpropyl 2-methylprop-2-enoate |
Specifications
| 97-86-9 | |
| 155°C | |
| MFCD00008931,MFCD00084435 | |
| 2283 | |
| RUMACXVDVNRZJZ-UHFFFAOYSA-N | |
| 2-methylpropyl 2-methylprop-2-enoate | |
| 7352 | |
| ≥98.0% (GC) | |
| Isobutyl Methacrylate (stabilized with HQ) |
| Colorless | |
| C8H14O2 | |
| 25 mL | |
| isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove | |
| CC(C)COC(=O)C(C)=C | |
| 142.20 | |
| 142.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1039
RTECSNumber : OZ4900000
TSCA : Yes