missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Methacrylate (stabilized with HQ) 98.0+%, TCI America™

Catalog No. M013225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M013225ML 25 mL
M0132500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M013225ML Supplier TCI America Supplier No. M013225ML

Chemical Identifiers

CAS 97-86-9
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00008931,MFCD00084435
InChI Key RUMACXVDVNRZJZ-UHFFFAOYSA-N
Synonym isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove
PubChem CID 7352
IUPAC Name 2-methylpropyl 2-methylprop-2-enoate
SMILES CC(C)COC(=O)C(C)=C

Specifications

Color Colorless
Boiling Point 155°C
Quantity 25 mL
UN Number 2283
Formula Weight 142.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Methacrylate (stabilized with HQ)
EINECSNumber (2)-1039
RTECSNumber OZ4900000
TSCA Yes