missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl Methacrylate Polymer, TCI America™
$33.56 - $216.24
Chemical Identifiers
| CAS | 9011-15-8 |
|---|---|
| Molecular Formula | C8H14O2 |
| MDL Number | MFCD00008931 |
| Synonym | Methacrylic Acid Isobutyl Ester Polymer |
Chemical Identifiers
| 9011-15-8 | |
| C8H14O2 | |
| MFCD00008931 | |
| Methacrylic Acid Isobutyl Ester Polymer |
Specifications
| White | |
| 9011-15-8 | |
| MFCD00008931 | |
| 25 g |
| Crystal-Powder at 20°C | |
| C8H14O2 | |
| Methacrylic Acid Isobutyl Ester Polymer | |
| Isobutyl Methacrylate Polymer |
Safety and Handling
EINECSNumber : (6)-0524
TSCA : Yes