missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl Methacrylate Polymer, TCI America™
Supplier: TCI America M008625G
Specifications
| White | |
| Crystal-Powder at 20°C | |
| C8H14O2 | |
| Methacrylic Acid Isobutyl Ester Polymer |
| Isobutyl Methacrylate Polymer | |
| 9011-15-8 | |
| MFCD00008931 | |
| 25 g |
Chemical Identifiers
| 9011-15-8 | |
| C8H14O2 | |
| MFCD00008931 | |
| Methacrylic Acid Isobutyl Ester Polymer |
Safety and Handling
EINECSNumber : (6)-0524
TSCA : Yes