missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl methacrylate, 99%, stabilized

Catalog No. AC412680010
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412680010 1 kg Glass Bottle
AC412680025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC412680010 Supplier Thermo Scientific Chemicals Supplier No. 412680010

CAS: 97-86-9 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-86-9
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00008931,MFCD00084435
InChI Key RUMACXVDVNRZJZ-UHFFFAOYSA-N
Synonym isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove
PubChem CID 7352
IUPAC Name 2-methylpropyl 2-methylprop-2-enoate
SMILES CC(C)COC(=O)C(C)=C

Specifications

Acidity 0.005% max. (as methacrylic acid)
Melting Point -37.0°C
Color Colorless
Density 0.8800g/mL
Boiling Point 155.0°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4190 to 1.4210
Linear Formula CH2=C(CH3)COOCH2CH(CH3)2
Quantity 1 kg
Specific Gravity 0.88
Solubility Information Solubility in water: slightly soluble
Stabilizer 15 to 20ppm MEHQ
Formula Weight 142.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isobutyl methacrylate, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Flammable liquid and
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 202-613-

RUO – Research Use Only