missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl methacrylate, 99.5+%, stab. with 10ppm 4-methoxyphenol

CAS: 97-86-9 | C8H14O2 | 142.20 g/mol

$62.67 - $471.34

Chemical Identifiers

CAS 97-86-9
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00008931,MFCD00084435
InChI Key RUMACXVDVNRZJZ-UHFFFAOYSA-N
Synonym isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove
PubChem CID 7352
IUPAC Name 2-methylpropyl 2-methylprop-2-enoate
SMILES CC(C)COC(=O)C(C)=C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA41948AK
View Documents
Thermo Scientific Chemicals
041948AK
250 mL N/A
Only null left
 
AA41948K2
View Documents
Thermo Scientific Chemicals
041948K2
1 L N/A
Only null left
 
AA41948K7
View Documents
Thermo Scientific Chemicals
041948K7
4 L N/A
Only null left
 
Description

Description

Plastic industry: used in the manufacture of containing active hydroxyl acrylic resin. Coatings with epoxy resin, diisocyanate and melamine formaldehyde resin configuration used for preparing a two-component coating. In oil industry as additives for lubricating oil washing. In electronics industry used as electron microscope to take off the water. Textile industry used in the manufacture of adhesive fabric. Used as chemical reagents in analytical chemistry. In addition, also used for water immiscible embedding agent, used in the synthesis of medical polymer materials, thermosetting coatings and adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

97-86-9
142.20
RUMACXVDVNRZJZ-UHFFFAOYSA-N
7352
CC(C)COC(=O)C(C)=C
C8H14O2
MFCD00008931,MFCD00084435
isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove
2-methylpropyl 2-methylprop-2-enoate

Specifications

97-86-9
155°C
C8H14O2
250 mL
1747595
Soluble in water at 20°C,2g/L
CC(C)COC(=O)C(C)=C
142.20
142.2
Liquid
0.889 g/mL
46°C (115°F)
MFCD00008931,MFCD00084435
UN2283
isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove
RUMACXVDVNRZJZ-UHFFFAOYSA-N
2-methylpropyl 2-methylprop-2-enoate
7352
≥99.5%
Isobutyl methacrylate, Stabilized with 10ppm 4-methoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H317-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c

H226-H315-H317-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOBUTYL METHACRYLATE, STABILIZED

EINECSNumber : 202-613-0

RTECSNumber : OZ4900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only