missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Laurate 96.0+%, TCI America™

Catalog No. p-7131813
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
L008425ML 25 mL
L0084500ML 500 mL
2 options

Catalog No. L008425ML

Supplier: TCI America L008425ML

Only null left

Chemical Identifiers

CAS 37811-72-6
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00048424
InChI Key SZFOMJOELAAHAF-UHFFFAOYSA-N
Synonym isobutyl laurate, lauric acid isobutyl ester, isobutyl dodecanoate, dodecanoic acid, 2-methylpropyl ester, isobutyl=laurate, acmc-1afre
PubChem CID 101338
IUPAC Name 2-methylpropyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(C)C

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 256.43
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Laurate
EINECSNumber (2)-0798
TSCA Yes