accessibility menu, dialog, popup

UserName

H3COOCH3CH3

CAS RN 37811-72-6

IUPAC Name: 2-methylpropyl dodecanoate
Synonyms: 2-methylpropyl dodecanoate
Molecular Weight (g/mol): 256.43
Molecular Formula: C16H32O2
InChi Key: SZFOMJOELAAHAF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OCC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results