missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl Laurate 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 500 mL |
| Formula Weight | 256.43 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Isobutyl Laurate |
Safety and Handling
| EINECSNumber | (2)-0798 |
| TSCA | Yes |