missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Laurate 96.0+%, TCI America™

Catalog No. L008425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L008425ML 25 mL
L0084500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L008425ML Supplier TCI America Supplier No. L008425ML

Chemical Identifiers

CAS 37811-72-6
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00048424
InChI Key SZFOMJOELAAHAF-UHFFFAOYSA-N
Synonym isobutyl laurate, lauric acid isobutyl ester, isobutyl dodecanoate, dodecanoic acid, 2-methylpropyl ester, isobutyl=laurate, acmc-1afre
PubChem CID 101338
IUPAC Name 2-methylpropyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(C)C

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 256.43
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Laurate
EINECSNumber (2)-0798
TSCA Yes