missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Isobutyrate 98.0+%, TCI America™

Catalog No. I0316500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0316500ML 500 mL
I031625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0316500ML Supplier TCI America Supplier No. I0316500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 97-85-8
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00008916
InChI Key RXGUIWHIADMCFC-UHFFFAOYSA-N
Synonym isobutyl isobutyrate, isobutyl 2-methylpropanoate, isobutyl isobutanoate, isobutyric acid, isobutyl ester, propanoic acid, 2-methyl-, 2-methylpropyl ester, 2-methylpropyl 2-methylpropionate, 2-methylpropyl isobutyrate, isobutylester kyseliny isomaselne, fema number 2189, 2-methyl-1-propyl 2-methylpropanoate
PubChem CID 7351
IUPAC Name 2-methylpropyl 2-methylpropanoate
SMILES CC(C)COC(=O)C(C)C

Specifications

Color Colorless
Boiling Point 147°C
Quantity 500 mL
UN Number 2528
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Isobutyrate
RTECSNumber NQ5250000
TSCA Yes