accessibility menu, dialog, popup

UserName

H3CCH3OOCH3CH3

CAS RN 97-85-8

IUPAC Name: 2-methylpropyl 2-methylpropanoate
Synonyms: isobutyl isobutyrate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: RXGUIWHIADMCFC-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)C(C)C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)