missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl isobutyrate, 98%

CAS: 97-85-8 | C8H16O2 | 144.21 g/mol

Supplier:  Thermo Scientific Chemicals L04655AU

 View more versions of this product

Catalog No. AAL04655AU


Only null left
Explore available promotions

Description

Description

Isobutyl isobutyrate is used principally as a solvent, especially for nitrocellulose lacquers and thinners as well as coatings for plastic substrates, and high-solids coatings. The majority of end uses are in coatings (automotive, industrial, wood furniture, and graphic arts).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyl isobutyrate is used principally as a solvent, especially for nitrocellulose lacquers and thinners as well as coatings for plastic substrates, and high-solids coatings. The majority of end uses are in coatings (automotive, industrial, wood furniture, and graphic arts).

Solubility
Soluble in water (1 g/L at 20oc).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P374-P380-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOBUTYL ISOBUTYRATE

EINECSNumber : 202-612-5

RTECSNumber : NQ5250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only