missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Formate 95.0+%, TCI America™

Catalog No. F0055500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0055500ML 500 mL
F005525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0055500ML Supplier TCI America Supplier No. F0055500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 542-55-2
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00003293
InChI Key AVMSWPWPYJVYKY-UHFFFAOYSA-N
Synonym isobutyl formate, isobutyl methanoate, tetryl formate, formic acid, 2-methylpropyl ester, iso-butyl formate, formic acid, isobutyl ester, isobutyl methanote, 2-methyl-1-propyl formate, isobutyl formate natural, isobutylester kyseliny mravenci
PubChem CID 10957
IUPAC Name 2-methylpropyl formate
SMILES CC(C)COC=O

Specifications

Color Yellow
Boiling Point 98°C
Quantity 500 mL
UN Number 2393
Formula Weight 102.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Formate
EINECSNumber (2)-0684
RTECSNumber LQ8650000
TSCA Yes