accessibility menu, dialog, popup

UserName

H3CCH3OO

CAS RN 542-55-2

IUPAC Name: 2-methylpropyl formate
Synonyms: isobutyl formate
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: AVMSWPWPYJVYKY-UHFFFAOYSA-N
SMILES: CC(C)COC=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results