missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl Benzoate 99.0+%, TCI America™
$65.24 - $333.67
Chemical Identifiers
| CAS | 120-50-3 |
|---|---|
| Molecular Formula | C11H14O2 |
| Molecular Weight (g/mol) | 178.23 |
| MDL Number | MFCD00048344 |
| InChI Key | KYZHGEFMXZOSJN-UHFFFAOYSA-N |
| Synonym | isobutyl benzoate, benzoic acid, isobutyl ester, benzoic acid, 2-methylpropyl ester, unii-kq6xz9wjii, isobutyl benzoate natural, fema no. 2185, kq6xz9wjii, benzoic acid isobutyl ester, benzoic acid, isobutyl ester 6ci,7ci,8ci, iso-butyl benzoate |
| PubChem CID | 61048 |
| ChEBI | CHEBI:87500 |
| IUPAC Name | 2-methylpropyl benzoate |
| SMILES | CC(C)COC(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 120-50-3 | |
| 178.23 | |
| KYZHGEFMXZOSJN-UHFFFAOYSA-N | |
| 61048 | |
| 2-methylpropyl benzoate |
| C11H14O2 | |
| MFCD00048344 | |
| isobutyl benzoate, benzoic acid, isobutyl ester, benzoic acid, 2-methylpropyl ester, unii-kq6xz9wjii, isobutyl benzoate natural, fema no. 2185, kq6xz9wjii, benzoic acid isobutyl ester, benzoic acid, isobutyl ester 6ci,7ci,8ci, iso-butyl benzoate | |
| CHEBI:87500 | |
| CC(C)COC(=O)C1=CC=CC=C1 |
Specifications
| 120-50-3 | |
| 237°C | |
| MFCD00048344 | |
| isobutyl benzoate, benzoic acid, isobutyl ester, benzoic acid, 2-methylpropyl ester, unii-kq6xz9wjii, isobutyl benzoate natural, fema no. 2185, kq6xz9wjii, benzoic acid isobutyl ester, benzoic acid, isobutyl ester 6ci,7ci,8ci, iso-butyl benzoate | |
| CC(C)COC(=O)C1=CC=CC=C1 | |
| 178.23 | |
| CHEBI:87500 | |
| ≥99.0% (GC) | |
| Isobutyl Benzoate |
| Colorless | |
| C11H14O2 | |
| 25 g | |
| KYZHGEFMXZOSJN-UHFFFAOYSA-N | |
| 2-methylpropyl benzoate | |
| 61048 | |
| 178.23 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-1356
RTECSNumber : DH4185000
TSCA : Yes