missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Alcohol, 99.0% min, ACS, MilliporeSigma™

Catalog No. p-4382134
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MBX17911 500 mL
MBX17915 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MBX17911 Supplier Technidata Supplier No. BX17911
Only null left

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Color Colorless
Density 0.802g/cm3 (20°C)
Explosion Limit 1.5 to 12%(V)
Ignition Point 430°C
Assay Percent Range 99.0% min.
Linear Formula (CH3)2CHCH2OH
Quantity 500 mL
Vapor Pressure 12 hPa (20°C)
Formula Weight 74.12
Grade ACS/EP/Reagent
Physical Form Liquid
Chemical Name or Material Isobutyl Alcohol, For ACS analysis
Show More Show Less
Recommended Storage Store without restrictions