missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl acetate, Analytical Standard, >99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193443
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020279 1 mL
111018804 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020279 Supplier Merck Emd Millipore Supplier No. 948231MLF

Isobutyl acetate is a chemical compound, commonly used as a solvent and flavoring agent in food products.

Specifications

CAS 110-19-0
Vapor Pressure 15 mmHg (20°C)
Vapor Density >4 (vs. air)
Density 0.867 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3COOCH2CH(CH3)2
MDL Number MFCD00008932
Melting Point -99°C
Boiling Point 115°C to 117°C
Synonym Acetic acid isobutyl ester
Quantity 1 mL
Formula Weight 116.16
Percent Purity >99.8% (GC)
Grade Analytical Standard
Show More Show Less