missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl 4-Hydroxybenzoate 99.0+%, TCI America™

Catalog No. p-7130565
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H049325G 25 g
H0493100G 100 g
H0493500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H049325G Supplier TCI America Supplier No. H049325G
Only null left

Chemical Identifiers

CAS 2-3-4247
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00020167
InChI Key XPJVKCRENWUEJH-UHFFFAOYSA-N
Synonym isobutyl 4-hydroxybenzoate, isobutylparaben, isobutyl p-hydroxybenzoate, isobutyl paraben, isobutyl-4-hydroxybenzoate, isobutyl parahydroxybenzoate, 4-hydroxybenzoic acid isobutyl ester, benzoic acid, 4-hydroxy-, 2-methylpropyl ester, p-oxybenzoesaureisobutylester, unii-0qqj25x58g
PubChem CID 20240
IUPAC Name 2-methylpropyl 4-hydroxybenzoate
SMILES CC(C)COC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 76°C
Color White
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isobutyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
RTECSNumber DH2247000
TSCA No