missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl 4-hydroxybenzoate, 98%

Catalog No. AAL1758706
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1758706 5 g
AAL1758718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1758706 Supplier Thermo Scientific Chemicals Supplier No. L1758706

CAS: 4247-02-3 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Isobutyl 4-hydroxybenzoate is used as an anti-microbial agent. It is also used as antiseptics for cosmetic field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2-3-4247
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00020167
InChI Key XPJVKCRENWUEJH-UHFFFAOYSA-N
Synonym isobutyl 4-hydroxybenzoate, isobutylparaben, isobutyl p-hydroxybenzoate, isobutyl paraben, isobutyl-4-hydroxybenzoate, isobutyl parahydroxybenzoate, 4-hydroxybenzoic acid isobutyl ester, benzoic acid, 4-hydroxy-, 2-methylpropyl ester, p-oxybenzoesaureisobutylester, unii-0qqj25x58g
PubChem CID 20240
IUPAC Name 2-methylpropyl 4-hydroxybenzoate
SMILES CC(C)COC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 74°C to 78°C
Quantity 5 g
Beilstein 2642305
Solubility Information Insoluble in water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material Isobutyl 4-hydroxybenzoate
EINECSNumber 224-208-8
RTECSNumber DH2247000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only