Learn More
Isobutanol, ACS, 99+%
Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.001%, Solubility in water P.T., Titratable acid 0.0005meq/g, H2O 0.1%, Carbonyl compounds 0.01% (butyraldehyde) and 0.02% (2-butanone) | CAS: 78-83-1 | C4H10O | 74.123 g/mol
Supplier: Thermo Scientific Chemicals 036643K7
Description
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful for the production of renewable chemicals and fuels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Isobutanol | |
| For analysis ACS | |
| Colorless | |
| 0.802 g/cm3 | |
| 4 L | |
| 390°C (734°F) | |
| Liquid | |
| >99% | |
| MFCD00004740 | |
| 1730878 | |
| Miscible with alcohol and diethyl ether. Partially miscible with water. | |
| CC(C)CO | |
| 74.123 | |
| CHEBI:46645 | |
| ACS |
| °C to 108°C | |
| 78-83-1 | |
| Colorless liquid | |
| 107°C to 108°C | |
| 28°C (82°F) | |
| Sweet | |
| 1.396 | |
| C4H10O | |
| 4-40 hPa (3-30mm Hg) at 20°C | |
| 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
| ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
| 2-methylpropan-1-ol | |
| 6560 | |
| 74.12 |
Chemical Identifiers
| 78-83-1 | |
| 74.123 | |
| ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
| 6560 | |
| 2-methylpropan-1-ol |
| C4H10O | |
| MFCD00004740 | |
| 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
| CHEBI:46645 | |
| CC(C)CO |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P332+P313-P363-P370+P378q-P501c
H226-H315-H318-H335-H336
DOTInformation : Hazard Class: 3, Packing Group: III
EINECSNumber : 201-148-0
RTECSNumber : NP9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only