missing translation for 'onlineSavingsMsg'
Learn More

Isobornyl acrylate, tech. 85%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

$134.88 - $134.88

Chemical Identifiers

CAS 5888-33-5
Molecular Formula C13H20O2
Molecular Weight (g/mol) 208.30
MDL Number MFCD00080424
InChI Key PSGCQDPCAWOCSH-BOURZNODSA-N
Synonym isobornyl acrylate, acrylic acid isobornyl ester, 1,7,7-trimethyltricyclo 2.2.1 hepten-2-yl-2-propenoate
PubChem CID 122130333
IUPAC Name [(3R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] prop-2-enoate
SMILES CC1(C)[C@H]2CC[C@]1(C)C(C2)OC(=O)C=C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0997730
View Documents
Thermo Scientific Chemicals
L0997730
250 g
Each for $134.88
Only null left
 
Description

Description

It is used as UVcuring EBcuring adhesive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as UVcuring EBcuring adhesive.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

5888-33-5
208.30
PSGCQDPCAWOCSH-BOURZNODSA-N
122130333
CC1(C)[C@H]2CC[C@]1(C)C(C2)OC(=O)C=C
C13H20O2
MFCD00080424
isobornyl acrylate, acrylic acid isobornyl ester, 1,7,7-trimethyltricyclo 2.2.1 hepten-2-yl-2-propenoate
[(3R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] prop-2-enoate

Specifications

5888-33-5
0.97
108°C (226°F)
C13H20O2
MFCD00080424
UN3082
Light sensitive
Difficult to mix in water.
CC1(C)[C@H]2CC[C@]1(C)C(C2)OC(=O)C=C
208.30
208.3
Technical
<-35°C
104°C to 106°C (4.5 mmHg)
Characteristic
1.4795
250 g
2254825
isobornyl acrylate, acrylic acid isobornyl ester, 1,7,7-trimethyltricyclo 2.2.1 hepten-2-yl-2-propenoate
PSGCQDPCAWOCSH-BOURZNODSA-N
[(3R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] prop-2-enoate
122130333
85%
Isobornyl acrylate, stabilized with 100ppm 4-methoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 227-561-6

RTECSNumber : UD3940000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only