missing translation for 'onlineSavingsMsg'
Learn More

Isobornyl acrylate, tech. 85%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0997730

 View more versions of this product

Catalog No. AAL0997730

Only null left
Explore available promotions

Description

Description

It is used as UVcuring EBcuring adhesive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as UVcuring EBcuring adhesive.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Specifications

Isobornyl acrylate
5888-33-5
0.97
108°C (226°F)
C13H20O2
MFCD00080424
UN3082
Light sensitive
Difficult to mix in water.
CC1(C)[C@H]2CC[C@]1(C)C(C2)OC(=O)C=C
208.30
208.3
Technical
stabilized with 100ppm 4-methoxyphenol
<-35°C
104°C to 106°C (4.5 mmHg)
Characteristic
1.4795
250 g
2254825
isobornyl acrylate, acrylic acid isobornyl ester, 1,7,7-trimethyltricyclo 2.2.1 hepten-2-yl-2-propenoate
PSGCQDPCAWOCSH-BOURZNODSA-N
[(3R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] prop-2-enoate
122130333
85%

Chemical Identifiers

5888-33-5
208.30
PSGCQDPCAWOCSH-BOURZNODSA-N
122130333
CC1(C)[C@H]2CC[C@]1(C)C(C2)OC(=O)C=C
C13H20O2
MFCD00080424
isobornyl acrylate, acrylic acid isobornyl ester, 1,7,7-trimethyltricyclo 2.2.1 hepten-2-yl-2-propenoate
[(3R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] prop-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 227-561-6

RTECSNumber : UD3940000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only