missing translation for 'onlineSavingsMsg'
Learn More

Isobornyl acetate, 94%

Catalog No. AAL11748AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL11748AE 100 mL
AAL11748AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL11748AE Supplier Thermo Scientific Chemicals Supplier No. L11748AE

CAS: 125-12-2 | C12H20O2 | 196.29 g/mol

Isobornyl acetate is one of the most important chemicals used in the perfumery industry. It is used in toiletries and soaps as a flavoring agent and antiseptics. One of main applications is as an intermediate to produce camphor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobornyl acetate is one of the most important chemicals used in the perfumery industry. It is used in toiletries and soaps as a flavoring agent and antiseptics. One of main applications is as an intermediate to produce camphor.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 125-12-2
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00867808,MFCD00135943
InChI Key KGEKLUUHTZCSIP-JFGNBEQYSA-N
Synonym 1s,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl acetate
PubChem CID 6950273
IUPAC Name [(1S,3S,4S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
SMILES CC(=O)O[C@H]1C[C@H]2CC[C@@]1(C)C2(C)C

Specifications

Density 0.99
Boiling Point 220°C to 224°C
Flash Point 89°C (192°F)
Refractive Index 1.465
Quantity 100 mL
Beilstein 3197572
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 196.29
Percent Purity 94%
Chemical Name or Material Isobornyl acetate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 204-727-6
RTECSNumber NP7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only