missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Isoborneol 90.0+%, TCI America™

Catalog No. p-7131304
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I027525G 25 g
I0275500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I027525G Supplier TCI America Supplier No. I027525G

Chemical Identifiers

CAS 124-76-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00074821
InChI Key DTGKSKDOIYIVQL-SZBHIRRCSA-N
Synonym dl-isoborneol
PubChem CID 126961757
IUPAC Name (1R,3R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
SMILES CC1(C2CCC1(C(C2)O)C)C

Specifications

Melting Point 211°C
Color White
Quantity 25 g
Formula Weight 154.25
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-Isoborneol
EINECSNumber (4)-0607
RTECSNumber NP7300000
TSCA Yes