CAS RN 124-76-5
IUPAC Name:
(1R,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol
Synonyms:
isoborneol
Molecular Weight (g/mol):
154.25
Molecular Formula:
C10H18O
InChi Key:
DTGKSKDOIYIVQL-XKSSXDPKSA-N
SMILES:
CC1(C)[C@H]2CC[C@@]1(C)[C@H](O)C2
1
–
2
of
2
results
1
DL-Isoborneol, 93%, tech.
CAS: 124-76-5 Molecular Formula: C10H18O Molecular Weight (g/mol): 154.25 MDL Number: MFCD00074821 InChI Key: DTGKSKDOIYIVQL-SZBHIRRCSA-N Synonym: dl-isoborneol PubChem CID: 126961757 IUPAC Name: (1R,3R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol SMILES: CC1(C2CCC1(C(C2)O)C)C
2
(+/-)-Isoborneol 90.0+%, TCI America™
CAS: 124-76-5 Molecular Formula: C10H18O Molecular Weight (g/mol): 154.253 MDL Number: MFCD00074821 InChI Key: DTGKSKDOIYIVQL-SZBHIRRCSA-N Synonym: dl-isoborneol PubChem CID: 126961757 IUPAC Name: (1R,3R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol SMILES: CC1(C2CCC1(C(C2)O)C)C