missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoamyl Mercaptan 94.0+%, TCI America™
$117.43 - $671.06
Chemical Identifiers
| CAS | 541-31-1 |
|---|---|
| Molecular Formula | C5H12S |
| Molecular Weight (g/mol) | 104.211 |
| MDL Number | MFCD00004899 |
| InChI Key | GIJGXNFNUUFEGH-UHFFFAOYSA-N |
| Synonym | isoamyl mercaptan, 3-methyl-1-butanethiol, 1-butanethiol, 3-methyl, isopentyl mercaptan, 3-methylbutanethiol, isoamylthiol, isopentanethiol, isoamylmercaptan, unii-mmk4sun45e, isoamyl sulfhydrate |
| PubChem CID | 10925 |
| IUPAC Name | 3-methylbutane-1-thiol |
| SMILES | CC(C)CCS |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I014925ML
|
TCI America
I014925ML |
25 mL |
Each for $117.43
|
|
|||||
|
I0149250ML
|
TCI America
I0149250ML |
250 mL |
Each for $671.06
|
|
|||||
Chemical Identifiers
| 541-31-1 | |
| 104.211 | |
| GIJGXNFNUUFEGH-UHFFFAOYSA-N | |
| 10925 | |
| CC(C)CCS |
| C5H12S | |
| MFCD00004899 | |
| isoamyl mercaptan, 3-methyl-1-butanethiol, 1-butanethiol, 3-methyl, isopentyl mercaptan, 3-methylbutanethiol, isoamylthiol, isopentanethiol, isoamylmercaptan, unii-mmk4sun45e, isoamyl sulfhydrate | |
| 3-methylbutane-1-thiol |
Specifications
| 541-31-1 | |
| 120°C | |
| MFCD00004899 | |
| 3336 | |
| GIJGXNFNUUFEGH-UHFFFAOYSA-N | |
| 3-methylbutane-1-thiol | |
| 10925 | |
| ≥94.0% (T) | |
| Isoamyl Mercaptan |
| Colorless | |
| C5H12S | |
| 25 mL | |
| isoamyl mercaptan, 3-methyl-1-butanethiol, 1-butanethiol, 3-methyl, isopentyl mercaptan, 3-methylbutanethiol, isoamylthiol, isopentanethiol, isoamylmercaptan, unii-mmk4sun45e, isoamyl sulfhydrate | |
| CC(C)CCS | |
| 104.211 | |
| 104.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0464
TSCA : Yes