missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Mercaptan 94.0+%, TCI America™

Supplier:  TCI America I0149250ML

 View more versions of this product

Catalog No. I0149250ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Isoamyl Mercaptan
Colorless
C5H12S
250 mL
isoamyl mercaptan, 3-methyl-1-butanethiol, 1-butanethiol, 3-methyl, isopentyl mercaptan, 3-methylbutanethiol, isoamylthiol, isopentanethiol, isoamylmercaptan, unii-mmk4sun45e, isoamyl sulfhydrate
CC(C)CCS
104.211
104.21
Liquid
541-31-1
120°C
MFCD00004899
3336
GIJGXNFNUUFEGH-UHFFFAOYSA-N
3-methylbutane-1-thiol
10925
≥94.0% (T)

Chemical Identifiers

541-31-1
104.211
GIJGXNFNUUFEGH-UHFFFAOYSA-N
10925
CC(C)CCS
C5H12S
MFCD00004899
isoamyl mercaptan, 3-methyl-1-butanethiol, 1-butanethiol, 3-methyl, isopentyl mercaptan, 3-methylbutanethiol, isoamylthiol, isopentanethiol, isoamylmercaptan, unii-mmk4sun45e, isoamyl sulfhydrate
3-methylbutane-1-thiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0464

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions