missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Laurate 97.0+%, TCI America™

Catalog No. p-7131771
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L001425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L001425ML Supplier TCI America Supplier No. L001425ML

Chemical Identifiers

CAS 6309-51-9
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00048429
InChI Key FVKRIDSRWFEQME-UHFFFAOYSA-N
Synonym isoamyl laurate, isopentyl laurate, dodecanoic acid, 3-methylbutyl ester, lauric acid, isopentyl ester, isoamyl dodecanoate, isopentyl dodecanoate, lauric acid isoamyl ester, isopentyl dodecylate, unii-m1slx00m3m, iso-amyl n-dodecanoate
PubChem CID 61386
ChEBI CHEBI:87343
IUPAC Name 3-methylbutyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCC(C)C

Specifications

Color Colorless
Boiling Point 170°C
Quantity 25 mL
Formula Weight 270.46
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Laurate
EINECSNumber (2)-0798
TSCA Yes