CAS RN 6309-51-9
IUPAC Name:
3-methylbutyl dodecanoate
Synonyms:
isoamyl laurate
Molecular Weight (g/mol):
270.46
Molecular Formula:
C17H34O2
InChi Key:
FVKRIDSRWFEQME-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC(=O)OCCC(C)C
1
–
1
of
1
results
1
Isoamyl Laurate 97.0+%, TCI America™
CAS: 6309-51-9 Molecular Formula: C17H34O2 Molecular Weight (g/mol): 270.457 MDL Number: MFCD00048429 InChI Key: FVKRIDSRWFEQME-UHFFFAOYSA-N Synonym: isoamyl laurate,isopentyl laurate,dodecanoic acid, 3-methylbutyl ester,lauric acid, isopentyl ester,isoamyl dodecanoate,isopentyl dodecanoate,lauric acid isoamyl ester,isopentyl dodecylate,unii-m1slx00m3m,iso-amyl n-dodecanoate PubChem CID: 61386 ChEBI: CHEBI:87343 IUPAC Name: 3-methylbutyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCC(C)C