missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoamyl Hexanoate 98.0+%, TCI America™
$46.20 - $138.61
Chemical Identifiers
| CAS | 2198-61-0 |
|---|---|
| Molecular Formula | C11H22O2 |
| Molecular Weight (g/mol) | 186.295 |
| MDL Number | MFCD00027280 |
| InChI Key | XVSZRAWFCDHCBP-UHFFFAOYSA-N |
| Synonym | Hexanoic Acid Isoamyl Ester, Isopentyl Hexanoate, Hexanoic Acid Isopentyl Ester |
| PubChem CID | 16617 |
| ChEBI | CHEBI:87542 |
| IUPAC Name | 3-methylbutyl hexanoate |
| SMILES | CCCCCC(=O)OCCC(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I089925ML
|
TCI America
I089925ML |
25 mL |
Each for $46.20
|
|
|||||
|
I0899100ML
|
TCI America
I0899100ML |
100 mL |
Each for $138.61
|
|
|||||
Chemical Identifiers
| 2198-61-0 | |
| 186.295 | |
| XVSZRAWFCDHCBP-UHFFFAOYSA-N | |
| 16617 | |
| 3-methylbutyl hexanoate |
| C11H22O2 | |
| MFCD00027280 | |
| Hexanoic Acid Isoamyl Ester, Isopentyl Hexanoate, Hexanoic Acid Isopentyl Ester | |
| CHEBI:87542 | |
| CCCCCC(=O)OCCC(C)C |
Specifications
| 2198-61-0 | |
| 98°C | |
| MFCD00027280 | |
| Hexanoic Acid Isoamyl Ester, Isopentyl Hexanoate, Hexanoic Acid Isopentyl Ester | |
| CCCCCC(=O)OCCC(C)C | |
| 186.295 | |
| CHEBI:87542 | |
| ≥98.0% (GC) | |
| Isoamyl Hexanoate |
| Colorless | |
| C11H22O2 | |
| 25 mL | |
| XVSZRAWFCDHCBP-UHFFFAOYSA-N | |
| 3-methylbutyl hexanoate | |
| 16617 | |
| 186.30 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0782
RTECSNumber : MO8389300
TSCA : Yes