accessibility menu, dialog, popup

UserName

CAS RN 2198-61-0

H3COOCH3CH3

CAS RN 2198-61-0

IUPAC Name: 3-methylbutyl hexanoate
Synonyms: isoamyl hexanoate
Molecular Weight (g/mol): 186.3
Molecular Formula: C11H22O2
InChi Key: XVSZRAWFCDHCBP-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OCCC(C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results