missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoamyl Ether (stabilized with BHT) 98.0+%, TCI America™
$42.36 - $213.69
Chemical Identifiers
| CAS | 544-01-4 |
|---|---|
| Molecular Formula | C10H22O |
| Molecular Weight (g/mol) | 158.285 |
| MDL Number | MFCD00008947 |
| InChI Key | AQZGPSLYZOOYQP-UHFFFAOYSA-N |
| Synonym | diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane |
| PubChem CID | 10989 |
| IUPAC Name | 3-methyl-1-(3-methylbutoxy)butane |
| SMILES | CC(C)CCOCCC(C)C |
Chemical Identifiers
| 544-01-4 | |
| 158.285 | |
| AQZGPSLYZOOYQP-UHFFFAOYSA-N | |
| 10989 | |
| CC(C)CCOCCC(C)C |
| C10H22O | |
| MFCD00008947 | |
| diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane | |
| 3-methyl-1-(3-methylbutoxy)butane |
Specifications
| 544-01-4 | |
| 173°C | |
| MFCD00008947 | |
| 3271 | |
| AQZGPSLYZOOYQP-UHFFFAOYSA-N | |
| 3-methyl-1-(3-methylbutoxy)butane | |
| 10989 | |
| ≥98.0% (GC) | |
| Isoamyl Ether (stabilized with BHT) |
| Colorless | |
| C10H22O | |
| 25 mL | |
| diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane | |
| CC(C)CCOCCC(C)C | |
| 158.285 | |
| 158.29 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0365
RTECSNumber : EK5433750
TSCA : Yes