accessibility menu, dialog, popup

UserName

H3CCH3OCH3CH3

CAS RN 544-01-4

IUPAC Name: 3-methyl-1-(3-methylbutoxy)butane
Synonyms: diisopentyl ether
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: AQZGPSLYZOOYQP-UHFFFAOYSA-N
SMILES: CC(C)CCOCCC(C)C
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)