missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoamyl Acetoacetate 97.0+%, TCI America™
$109.98 - $974.90
Chemical Identifiers
| CAS | 2308-18-1 |
|---|---|
| Molecular Formula | C9H16O3 |
| Molecular Weight (g/mol) | 172.224 |
| MDL Number | MFCD00059355 |
| InChI Key | XHRGPLDMNNGHCX-UHFFFAOYSA-N |
| Synonym | Acetoacetic Acid Isoamyl Ester, Isopentyl Acetoacetate |
| PubChem CID | 61296 |
| IUPAC Name | 3-methylbutyl 3-oxobutanoate |
| SMILES | CC(C)CCOC(=O)CC(=O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A081225ML
|
TCI America
A081225ML |
25 mL |
Each for $109.98
|
|
|||||
|
A0812500ML
|
TCI America
A0812500ML |
500 mL |
Each for $974.90
|
|
|||||
Chemical Identifiers
| 2308-18-1 | |
| 172.224 | |
| XHRGPLDMNNGHCX-UHFFFAOYSA-N | |
| 61296 | |
| CC(C)CCOC(=O)CC(=O)C |
| C9H16O3 | |
| MFCD00059355 | |
| Acetoacetic Acid Isoamyl Ester, Isopentyl Acetoacetate | |
| 3-methylbutyl 3-oxobutanoate |
Specifications
| 2308-18-1 | |
| 218°C | |
| MFCD00059355 | |
| Acetoacetic Acid Isoamyl Ester, Isopentyl Acetoacetate | |
| CC(C)CCOC(=O)CC(=O)C | |
| 172.224 | |
| 172.22 | |
| Liquid |
| Colorless | |
| C9H16O3 | |
| 25 mL | |
| XHRGPLDMNNGHCX-UHFFFAOYSA-N | |
| 3-methylbutyl 3-oxobutanoate | |
| 61296 | |
| ≥97.0% (GC) | |
| Isoamyl Acetoacetate |
Safety and Handling
EINECSNumber : (2)-1505
TSCA : No