accessibility menu, dialog, popup

UserName

CAS RN 2308-18-1

H3CCH3OOCH3O

CAS RN 2308-18-1

IUPAC Name: 3-methylbutyl 3-oxobutanoate
Synonyms: 3-methylbutyl 3-oxobutanoate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: XHRGPLDMNNGHCX-UHFFFAOYSA-N
SMILES: CC(C)CCOC(=O)CC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results