missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Acetate 98.0+%, TCI America™

Supplier:  TCI America A0264100ML

Catalog No. A0264100ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Isoamyl Acetate [for Spectrophotometry]
Colorless
C7H14O2
100 mL
isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate
CC(C)CCOC(C)=O
130.19
CHEBI:31725
≥98.0% (GC)
123-92-2
142°C
MFCD00008946
1104
MLFHJEHSLIIPHL-UHFFFAOYSA-N
3-methylbutyl acetate
31276
130.19
Liquid

Chemical Identifiers

123-92-2
130.19
MLFHJEHSLIIPHL-UHFFFAOYSA-N
31276
3-methylbutyl acetate
C7H14O2
MFCD00008946
isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate
CHEBI:31725
CC(C)CCOC(C)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0733

RTECSNumber : NS9800000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions