missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl 4-(Dimethylamino)benzoate 98.0+%, TCI America™

Catalog No. p-7131715
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I101325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I101325G Supplier TCI America Supplier No. I101325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21245-01-2
Molecular Formula C14H21NO2
Molecular Weight (g/mol) 235.327
MDL Number MFCD00059356
InChI Key OFSAUHSCHWRZKM-UHFFFAOYSA-N
Synonym isoamyl 4-dimethylamino benzoate, spectraban, 4-dimethylaminobenzoic acid isoamyl ester, 3-methylbutyl-4-dimethylaminobenzoate, isoamyl para-n,n-dimethylaminobenzoate, 3-methylbutyl 4-dimethylamino benzoate, benzoic acid, 4-dimethylamino-, 3-methylbutyl ester, 3-methylbutyl4-dimethylaminobenzoate, dsstox_cid_26580, dsstox_rid_81739
PubChem CID 88836
IUPAC Name 3-methylbutyl 4-(dimethylamino)benzoate
SMILES CC(C)CCOC(=O)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 155°C
Quantity 25 g
Formula Weight 235.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Isoamyl 4-(Dimethylamino)benzoate
EINECSNumber (3)-1436
TSCA No