accessibility menu, dialog, popup

UserName

CAS RN 21245-01-2

IUPAC Name: 3-methylbutyl 4-(dimethylamino)benzoate
Synonyms: padimate A
Molecular Weight (g/mol): 235.33
Molecular Formula: C14H21NO2
InChi Key: OFSAUHSCHWRZKM-UHFFFAOYSA-N
SMILES: CC(C)CCOC(=O)C1=CC=C(C=C1)N(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results