missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iso-Propyl Alcohol/Hexanes, 46/54 w/w, Reagents
Iso-Propyl Alcohol/Hexanes, 46/54 w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$244.60 - $5954.77
Chemical Identifiers
| CAS | 110-54-3 |
|---|---|
| Molecular Formula | C6H14 |
| Molecular Weight (g/mol) | 86.18 |
| MDL Number | MFCD02179311 |
| InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
| IUPAC Name | hexane |
| SMILES | CCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-334-930
|
Labchem Inc
CI171400200L |
200 L |
Each for $5,954.77
|
|
|||||
|
01-334-929
|
Labchem Inc
CI1714004C |
4 L |
Each for $244.60
|
|
|||||
Specifications
| 110-54-3,67-63-0 | |
| 200 L | |
| Liquid | |
| C6H14 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| hexane | |
| Reagent |
| Colorless | |
| Steel Drum | |
| 0.7164 | |
| MFCD02179311 | |
| CCCCCC | |
| 86.18 | |
| Iso-Propyl Alcohol/Hexanes, 46/54 w/w |