missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iso-Propyl Alcohol/Hexanes, 46/54 w/w, Reagents
Iso-Propyl Alcohol/Hexanes, 46/54 w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CI1714004C
Specifications
| Iso-Propyl Alcohol/Hexanes, 46/54 w/w | |
| Colorless | |
| Glass Bottle | |
| 0.7164 | |
| MFCD02179311 | |
| CCCCCC | |
| 86.18 |
| 110-54-3,67-63-0 | |
| 4 L | |
| Liquid | |
| C6H14 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| hexane | |
| Reagent |
Chemical Identifiers
| 110-54-3 | |
| 86.18 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| CCCCCC |
| C6H14 | |
| MFCD02179311 | |
| hexane |