missing translation for 'onlineSavingsMsg'
Learn More

Iso-Propyl Alcohol/Hexanes, 46/54 w/w, Reagents

Catalog No. 01334930
Change view
Click to view available options
Quantity:
4 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334930 200 L
01334929 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334930 Supplier Labchem Inc Supplier No. CI171400200L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Iso-Propyl Alcohol/Hexanes, 46/54 w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 110-54-3 , 67-63-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC

Specifications

Color Colorless
Quantity 200 L
Packaging Steel Drum
Physical Form Liquid
Specific Gravity 0.7164
Grade Reagent
Chemical Name or Material Iso-Propyl Alcohol/Hexanes, 46/54 w/w