missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iso-Propyl Alcohol ACS, MilliporeSigma™
$114.56 - $725.86
Chemical Identifiers
| CAS | 67-63-0 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | 60.096 |
| InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
| Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
| PubChem CID | 3776 |
| ChEBI | CHEBI:17824 |
| IUPAC Name | propan-2-ol |
| SMILES | CC(C)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
MPX18356
|
Technidata
PX18356 |
500 mL | Glass Bottle |
N/A
|
|
||||
|
MPX18359
|
Technidata
PX18359 |
1 L | Poly Bottle |
N/A
|
|
||||
|
MPX18355
|
Technidata
PX18355 |
4 L | Glass Bottle |
N/A
|
|
||||
|
MPX18353
|
Technidata
PX18353 |
20 L | Metal Can |
N/A
|
|
||||
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
Specifications
| 67-63-0 | |
| Glass Bottle | |
| CH3CHOHCH3 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 60.1 | |
| iso-Propyl Alcohol |
| 500 mL | |
| C3H8O | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| ACS Reagent |