missing translation for 'onlineSavingsMsg'
Learn More

iso-Propyl Alcohol ACS, MilliporeSigma™

Catalog No. MPX18359
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Glass Bottle
Metal Can
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX18359 1 L Poly Bottle
MPX18356 500 mL Glass Bottle
MPX18355 4 L Glass Bottle
MPX18353 20 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MPX18359 Supplier Technidata Supplier No. PX18359

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Quantity 1 L
Packaging Poly Bottle
Linear Formula CH3CHOHCH3
Formula Weight 60.1
Grade ACS
Chemical Name or Material iso-Propyl Alcohol