missing translation for 'onlineSavingsMsg'
Learn More

iso-Propyl Acetate, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P148420LT

 View more versions of this product

Catalog No. 18611115

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

108-21-4
Steel Can
0.98
1
20 L
Reagent

Chemical Identifiers

108-21-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.