missing translation for 'onlineSavingsMsg'
Learn More

Iso-Octane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0%, Solstice

Catalog No. 6000872
Change view
Click to view available options
Quantity:
204 L
Packaging:
Metal Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000872 204 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000872 Supplier Solstice Supplier No. PP362204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

CAS Max % 100.0000%
Density 0.69 g/cm3
Quantity 204 L
Flash Point −8°C
Packaging Metal Drum
Water 0.01% max.
UN Number UN1262
Beilstein 1696876
Boiling Point 99.24°C
Solubility Information 0.02% in Water at 20°C
Formula Weight 114.23
Percent Purity >99%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Iso-Octane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Octanes