missing translation for 'onlineSavingsMsg'
Learn More

Iso-Octane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%, Honeywell

Catalog No. p-7112055
Change view
Click to view available options
Quantity:
19 L
4 x 4 L
Packaging:
Glass Bottle
Metal Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000874 4 x 4 L Glass Bottle
6000873 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000874 Supplier Honeywell Inc Supplier No. LP3624
Only null left

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

CAS Max % 100.0000%
Density 0.69 g/cm3
Quantity 4 x 4 L
Flash Point −8°C
Packaging Glass Bottle
UN Number UN1262
Beilstein 1696876
Water 0.01% max.
Solubility Information 0.02% in Water at 20°C
Formula Weight 114.23
Percent Purity >99%
Grade ACS Reagent
Assay by GC Analysis
Chemical Name or Material Iso-Octane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Octanes