missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iso-Butyl Alcohol, ACS Reagent (2-Methyl-1-Propanol), Reagents
Specifications
Specifications
| Melting Point | -108°C |
| Color | Colorless |
| Quantity | 4 L |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.8016 |
| Boiling Point | 108°C |
| Grade | ACS |
| Chemical Name or Material | Iso-Butyl Alcohol, ACS Reagent (2-Methyl-1-Propanol) |