missing translation for 'onlineSavingsMsg'
Learn More

iso-Butyl Alcohol, ACS, 99%, Spectrum™ Chemical

Catalog No. 18611103
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611103 200 L Drum
18611105 100 mL Amber Glass Bottle
18611104 500 mL Amber Glass Bottle
18611100 1 L Amber Glass Bottle
18611101 4 L Amber Glass Bottle
18611102 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611103 Supplier Spectrum Chemical Mfg Cor Supplier No. B1186200LTBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78-83-1
Molecular Weight (g/mol) 74.12

Specifications

CAS Max % 1
Quantity 200 L
Packaging Drum
Residue after Evaporation 1e-05
Titratable Acid 0.0005 meq/g
Water 0.001
Boiling Point 108°C
Percent Purity 0.99
Grade Reagent
Chemical Name or Material Iso-Butyl Alcohol

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.